Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE4B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298954AF350
Description
UBE4B Polyclonal specifically detects UBE4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
UBE4B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
E4, homologous to yeast UFD2, Homozygously deleted in neuroblastoma 1, homozygously deleted in neuroblastoma-1, KIAA0684HDNB1, ubiquitin conjugation factor E4 B, Ubiquitin fusion degradation protein 2, ubiquitination factor E4B (homologous to yeast UFD2), ubiquitination factor E4B (UFD2 homolog, yeast), ubiquitin-fusion degradation protein 2, UBOX3, UFD2UFD2A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human UBE4B. | |
0.1 mL | |
Apoptosis | |
10277 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction