Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522023
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (90%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
Specifications
UBE4B | |
Polyclonal | |
Unconjugated | |
UBE4B | |
4930551I19Rik; 4933406G05Rik; AU014668; D4Bwg0973e; E4; HDNB1; homologous to yeast UFD2; homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; KIAA0684; mKIAA0684; RING-type E3 ubiquitin transferase E4 B; Ube4b; Ubiquitin conjugation factor E4 B; ubiquitin fusion degradation protein 2; ubiquitination factor E4B; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); ubiquitination factor E4B, UFD2 homolog; ubiquitin-fusion degradation protein 2; UBOX3; UFD2; Ufd2a; UFD2A-III/UBE4B-III splice isoform; Ufd2p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10277 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O95155 | |
UBE4B | |
Recombinant fragment corresponding to a region within amino acids 869 and 1179 of UBE4B (Uniprot ID#O95155). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction