Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581283 Supplier Invitrogen™ Supplier No. PA581283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human UBE4B.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE4B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene UBE4B
Gene Accession No. O95155
Gene Alias 4930551I19Rik; 4933406G05Rik; AU014668; D4Bwg0973e; E4; HDNB1; homologous to yeast UFD2; homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; KIAA0684; mKIAA0684; RING-type E3 ubiquitin transferase E4 B; Ube4b; Ubiquitin conjugation factor E4 B; ubiquitin fusion degradation protein 2; ubiquitination factor E4B; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); ubiquitination factor E4B, UFD2 homolog; ubiquitin-fusion degradation protein 2; UBOX3; UFD2; Ufd2a; UFD2A-III/UBE4B-III splice isoform; Ufd2p
Gene Symbols UBE4B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human UBE4B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.