Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE4B Recombinant Rabbit Monoclonal Antibody (1D7K5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542592
This item is not returnable.
View return policy
Description
Positive test controls include: PC-12, SKOV3, NIH/3T3, K-562, Mouse liver, Mouse brain, Rat liver, Rat brain. Immunogen sequence: MEELSADEIR RRRLARLAGG QTSQPTTPLT SPQRENPPGP PIAASAPGPS QSLGLNVHNM TPATSPIGAS GVAHRSQSSE GVSSLSSSPS NSLETQSQSL.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
Specifications
UBE4B | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95155, Q9ES00 | |
UBE4B | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBE4B (O95155). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
1D7K5 | |
Unconjugated | |
UBE4B | |
4930551I19Rik; 4933406G05Rik; AU014668; D4Bwg0973e; E4; HDNB1; homologous to yeast UFD2; homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; KIAA0684; mKIAA0684; RING-type E3 ubiquitin transferase E4 B; Ube4b; Ubiquitin conjugation factor E4 B; ubiquitin fusion degradation protein 2; ubiquitination factor E4B; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); ubiquitination factor E4B, UFD2 homolog; ubiquitin-fusion degradation protein 2; UBOX3; UFD2; Ufd2a; UFD2A-III/UBE4B-III splice isoform; Ufd2p | |
Rabbit | |
Affinity chromatography | |
RUO | |
10277, 298652, 63958 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction