Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE4B Recombinant Rabbit Monoclonal Antibody (JE55-76)
GREENER_CHOICE

Catalog No. PIMA542492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542492 Supplier Invitrogen™ Supplier No. MA542492
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: HepG2 cell lysate, NIH/3T3 cell lysate, PC-12 cell lysate, rat skeletal muscle tissue, human lung carcinoma tissue, N2A whole cell lysate. Subcellular Location: Nucleus, Cytoplasm.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE55-76
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UBE4B
Gene Accession No. O95155, Q9ES00
Gene Alias 4930551I19Rik; 4933406G05Rik; AU014668; D4Bwg0973e; E4; HDNB1; homologous to yeast UFD2; homozygously deleted in neuroblastoma 1; homozygously deleted in neuroblastoma-1; KIAA0684; mKIAA0684; RING-type E3 ubiquitin transferase E4 B; Ube4b; Ubiquitin conjugation factor E4 B; ubiquitin fusion degradation protein 2; ubiquitination factor E4B; ubiquitination factor E4B (homologous to yeast UFD2); ubiquitination factor E4B (UFD2 homolog, yeast); ubiquitination factor E4B, UFD2 homolog; ubiquitin-fusion degradation protein 2; UBOX3; UFD2; Ufd2a; UFD2A-III/UBE4B-III splice isoform; Ufd2p
Gene Symbols UBE4B
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal Human UBE4B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10277, 298652, 63958
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.