Learn More
Invitrogen™ UBF-1 Polyclonal Antibody

Description
Immunogen sequence: WKLLSQKEKD AYHKKCDQKK KDYEVELLRF LESLPEEEQQ RVLGEEKMLN INKKQATSPA SKKPAQEGGK GGSEKPKRPV SAMFIFSEEK RRQLQEERPE LSESELTRLL ARMWNDLSEK KK Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
Specifications
Specifications
| Antigen | UBF-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | UBTF |
| Gene Accession No. | P17480, P25976 |
| Gene Alias | 90-kDa nucleolus organizer region autoantigen; A930005G04Rik; Autoantigen NOR-90; NOR90; NOR-90; Nucleolar transcription factor 1; Tcfubf; transcription factor UBF; UBF; UBF1; UBF-1; UBF2; Ubtf; upstream binding transcription factor, RNA polymerase I; upstream-binding factor 1 |
| Gene Symbols | UBTF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.