Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBFD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106765 Supplier Invitrogen™ Supplier No. PA5106765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UBFD1.

May play a role as NF-kappa-B regulator. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBFD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UBFD1
Gene Accession No. O14562, Q78JW9
Gene Alias AI467302; D7Wsu105e; D7Wsu128e; RGD1306614; UBFD1; Ubiquitin domain-containing protein UBFD1; ubiquitin family domain containing 1; ubiquitin-binding protein homolog; UBPH
Gene Symbols UBFD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human UBFD1(Accession O14562), corresponding to amino acid residues K198-T248.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28018, 293454, 56061
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.