Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    UBIAD1 Antibody, Allophycocyanin, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100468APC
Description
UBIAD1 Polyclonal antibody specifically detects UBIAD1 in Human samples. It is validated for Western BlotSpecifications
| UBIAD1 | |
| Polyclonal | |
| Western Blot | |
| EC 2.5.1.-, RP4-796F18.1, SCCD, Schnyder crystalline corneal dystrophy, TERE1transitional epithelia response protein, Transitional epithelial response protein 1, UbiA prenyltransferase domain containing 1, ubiA prenyltransferase domain-containing protein 1 | |
| A synthetic peptide made to residues 32-45 [PEQDRLPQRSWRQK] of the 37 kDa human TERE1 protein. | |
| 0.1 mL | |
| Cancer | |
| 29914 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            