Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubidecarenone, USP Dietary Supplement, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18607632 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail

U1310, 303-98-0, C59H90O4

Spectrum™ Chemical Ubidecarenone, USP Dietary Supplement , Coenzyme Q-10, has not been approved for therapeutic use in the US, but it is available as a food supplement. All Spectrum Chemical USP Dietary Supplement products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 303-98-0
Molecular Formula C59H90O4
Molecular Weight (g/mol) 863.37
MDL Number MFCD00042919
InChI Key ACTIUHUUMQJHFO-UPTCCGCDSA-N
IUPAC Name 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaen-1-yl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CCC=C(C)C)=C(C)C1=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98 to 101%
Grade USP Dietary Supplement
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.