Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ubinuclein 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554071
Description
Immunogen sequence: SVTNQNVTPF GMLGGLVPVT MPFQFPLEIF GFGTDTAGVT TTSGSTSAAF HHSLTQNLLK GLQPGGAQHA ATLSHSPLPA HLQQAFHDGG QSKGDTKLPR KS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
Ubinuclein 1 (UBN1) is a ubiquitously expressed evolutionarily conserved protein which binds to proliferation-promoting genes that are repressed by formation of senescence-associated heterochromatin foci (SAHF). Ubinuclein 1 associates with various transcription factors and with histone methyltransferase activity, is indispensable for SAHF formation and appears to be a regulator of senescence. Although in most cells ubinuclein is localized to the nucleus, in cells forming tight junctions it is recruited to the cell adhesion complexes, dependently on the cell density.
Specifications
Ubinuclein 2 | |
Polyclonal | |
Unconjugated | |
UBN2 | |
2900060J04Rik; 6030408G03Rik; BC060116; D130059P03Rik; Kiaa2030; mKIAA2030; ubinuclein 1; ubinuclein 2; ubinuclein-2; UBN1; UBN2; VT; VT4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254048 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZU65 | |
UBN2 | |
Recombinant protein corresponding to Human Ubinuclein 1/2. Recombinant protein control fragment (Product #RP-92771). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction