Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ubinuclein 1/2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537942
Description
This antibody is predicted to react with mouse, rat, dog, pig, cow and zebrafish based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Ubinuclein 1 (UBN1) is a ubiquitously expressed evolutionarily conserved protein which binds to proliferation-promoting genes that are repressed by formation of senescence-associated heterochromatin foci (SAHF). Ubinuclein 1 associates with various transcription factors and with histone methyltransferase activity, is indispensable for SAHF formation and appears to be a regulator of senescence. Although in most cells ubinuclein is localized to the nucleus, in cells forming tight junctions it is recruited to the cell adhesion complexes, dependently on the cell density.
Specifications
Ubinuclein 1/2 | |
Polyclonal | |
Unconjugated | |
UBN2 | |
HIRA-binding protein; KIAA2030; Protein VT4; ubinuclein 1; ubinuclein 2; ubinuclein-1; Ubinuclein-2; ubiquitously expressed nuclear protein; UBN1; UBN2; VT; VT4 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
254048, 29855 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q6ZU65, Q9NPG3 | |
UBN1, UBN2 | |
Peptide with sequence C-DNSEAYDELVPASLT, from the internal region of UBN1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction