Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubinuclein 1 Monoclonal Antibody (UBN1-02)
GREENER_CHOICE

Catalog No. PIMA110458
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110458 Supplier Invitrogen™ Supplier No. MA110458
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes N-terminal part of ubinuclein 1 (UBN1), a widely expressed nuclear and adhesion complex protein. Western blotting analysis reveals UBN1 as a 165 kDa band.

Cellular senescence is a hallmark of tumor suppression and tissue aging. Senescent cells contain domains of heterochromatin, called senescence-associated heterochromatin foci (SAHF), that repress proliferation-promoting genes. The protein encoded by this gene binds to proliferation-promoting genes and is required for SAHF formation, enhancing methylation of histone H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubinuclein 1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone UBN1-02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene UBN1
Gene Accession No. Q9NPG3
Gene Alias 1110029L11Rik; 2610108L02Rik; AA673476; AW124741; HIRA-binding protein; Protein VT4; ubinuclein 1; ubinuclein-1; Ubiquitously expressed nuclear protein; Ubn1; VT; VT4
Gene Symbols UBN1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29855
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.