Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquilin 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203341
Description
Ubiquilin 1 Polyclonal specifically detects Ubiquilin 1 in Human, Mouse, Chinese Hamster, Hamster samples. It is validated for Western Blot.Specifications
Ubiquilin 1 | |
Polyclonal | |
Unconjugated | |
PBS with 1mg/ml BSA with 0.05% Sodium Azide | |
UBQLN1 | |
Synthetic Peptide: A(2) E S A E S G G P P G A Q D S A A(18) | |
100 μg | |
Primary | |
Ubiquilin 1 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
1 mg/mL | |
Western Blot 1 ug/ml | |
DA41FLJ90054, DSK2, hPLIC-1, PLIC1, PLIC-1UBQN, Protein linking IAP with cytoskeleton 1, ubiquilin 1, ubiquilin-1, XDRP1 | |
Rabbit | |
Affinity Purified | |
RUO | |
29979 | |
Human, Mouse, Chinese Hamster, Hamster | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction