Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquilin 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Ubiquilin 2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17975320
![]() |
Novus Biologicals
NBP17975320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179753
![]() |
Novus Biologicals
NBP179753 |
100 μL |
Each for $487.50
|
|
|||||
Description
Ubiquilin 2 Polyclonal specifically detects Ubiquilin 2 in Human samples. It is validated for Western Blot.Specifications
Ubiquilin 2 | |
Polyclonal | |
Rabbit | |
NP_038472 | |
29978 | |
Synthetic peptide directed towards the N terminal of human UBQLN2. Peptide sequence LKDQDTLIQHGIHDGLTVHLVIKSQNRPQGQSTQPSNAAGTNTTSASTPR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2 | |
UBQLN2 | |
IgG | |
66 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title