Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubiquilin 2 Monoclonal Antibody (6H9)

Mouse Monoclonal Antibody

$526.00 - $1800.00

Specifications

Antigen Ubiquilin 2
Clone 6H9
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA547367
View Documents
Invitrogen™
MA547367
500 μL
Each for $1,800.00
Only null left
Add to Cart
 
PIMA547466
View Documents
Invitrogen™
MA547466
100 μL
Each for $526.00
Only null left
Add to Cart
 
Description

Description

Ubiquilin 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC

This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases; and thus, are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to bind the ATPase domain of the Hsp70-like Stch protein.
Specifications

Specifications

Ubiquilin 2
1 mg/mL
Immunohistochemistry, Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 50% glycerol and 5 mM sodium azide
Q9QZM0, Q9UHD9
29978, 317396, 54609
Human ubiquilin 2 expressed in and purified from E. coli.
Antibody
6H9
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG1
Human, Mouse, Rat
UBQLN2
ALS15; CHAP1; DSK2; DSK2 homolog; HRIHFB2157; N4BP4; Nedd4 binding protein 4; Plic2; Plic-2; protein linking IAP with cytoskeleton 2; RGD1563566; ubiquilin 2; ubiquilin-2; Ubiquitin-like product Chap1/Dsk2; Ubiquitin-related protein; Ubqln2
UBQLN2
Primary
Protein A/G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.