Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubiquilin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104192 Supplier Invitrogen™ Supplier No. PA5104192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UBQLN2.

This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases; and thus, are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to bind the ATPase domain of the Hsp70-like Stch protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquilin 2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UBQLN2
Gene Accession No. Q9QZM0, Q9UHD9
Gene Alias ALS15; CHAP1; DSK2; DSK2 homolog; HRIHFB2157; N4BP4; Nedd4 binding protein 4; Plic2; Plic-2; protein linking IAP with cytoskeleton 2; RGD1563566; ubiquilin 2; ubiquilin-2; Ubiquitin-like product Chap1/Dsk2; Ubiquitin-related protein; Ubqln2
Gene Symbols UBQLN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human UBQLN2(Accession Q9UHD9), corresponding to amino acid residues S527-N577.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29978, 317396, 54609
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.