Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubiquilin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582249 Supplier Invitrogen™ Supplier No. PA582249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSAMSNPRAM QALMQIQQGL QTLATEAPGL IPSFTPGVGV GVLGTAIGPV GPVTPIGPIG PIVPFTPIGP IGPIGPTGPA APPGSTGSGG PTGPTVSSAA PSETTSPTSE SGPNQQFIQQ MVQALAGANA PQLPNPEVRF QQQLEQLN Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases; and thus, are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to bind the ATPase domain of the Hsp70-like Stch protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquilin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBQLN2
Gene Accession No. Q9UHD9
Gene Alias ALS15; CHAP1; DSK2; DSK2 homolog; HRIHFB2157; N4BP4; Nedd4 binding protein 4; Plic2; Plic-2; protein linking IAP with cytoskeleton 2; RGD1563566; ubiquilin 2; ubiquilin-2; Ubiquitin-like product Chap1/Dsk2; Ubiquitin-related protein; Ubqln2
Gene Symbols UBQLN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBQLN2. Recombinant protein control fragment (Product #RP-92271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.