Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Activating Enzyme E1Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501990846
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0846 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0846 Supplier Rockland Immunochemicals Supplier No. 200401888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is directed against human Ubiquitin Activating Enzyme E1 protein. A BLAST analysis was used to suggest that this antibody would react with Ubiquitin Activating Enzyme E1 protein from human (100%) rabbit (96%), mouse (95%), rat (95%) and dog (93%) based on a high degree of sequence homology. Cross reactivity against this protein from other sources has not been determined.

UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.

Specifications

Antigen Ubiquitin Activating Enzyme E1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene UBA1
Gene Accession No. P22314
Gene Alias A1S9; A1S9 protein; A1S9T; A1S9T and BN75 temperature sensitivity complementing; A1ST; AMCX1; CFAP124; EMBL:AAH85791.1, ECO:0000312; GXP1; POC20; POC20 centriolar protein homolog; Protein A1S9; RGD:1359327}; Sbx; SMAX2; testicular secretory protein Li 63; Uba1; uba1 {ECO:0000312; UBA1, ubiquitin-activating enzyme E1 homolog A; UBA1A; UBE1; Ube-1; Ube1ax; Ube1x; ubiquitin like modifier activating enzyme 1; Ubiquitin-activating enzyme E1; ubiquitin-activating enzyme E1 {ECO:0000250; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; Ubiquitin-like modifier-activating enzyme 1; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250; Ubiquitin-like modifier-activating enzyme 1 X; UniProtKB:Q02053}
Gene Symbols UBA1
Host Species Rabbit
Immunogen Anti-Ubiquitin Activating Enzyme E1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to full length Human Ubiquitin Activating Enzyme E1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.