Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin B Antibody (6C11-B3), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212882R
Description
Ubiquitin B Monoclonal specifically detects Ubiquitin B in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Ubiquitin B | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UBB | |
Native bovine ubiquitin, conjugated to KLH | |
200 μL | |
7314 | |
Human, Mouse, Rat, Bovine | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
6C11-B3 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 10kDa. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction