Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin B Antibody (UBB/1748), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308905MFV450
Description
Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Flow CytometrySpecifications
Ubiquitin B | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
UBB/1748 | |
Flow Cytometry | |
FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary) | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
7314 | |
Store at 4C in the dark. | |
IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction