Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin B Antibody (UBB/2122), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308906JF549
Description
Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Ubiquitin B | |
Monoclonal | |
Janelia Fluor 549 | |
FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary) | |
100 μg | |
Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
7314 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array | |
UBB/2122 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction