Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubiquitin B Monoclonal Antibody (3C12)
GREENER_CHOICE

Catalog No. PIMA517196
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517196 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517196 Supplier Invitrogen™ Supplier No. MA517196
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17196 targets UBB in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17196 immunogen is purified recombinant fragment of human UBB expressed in E. Coli. MA5-17196 detects UBB which has a predicted molecular weight of approximately 26kDa.

Ubiquitin is a highly conserved protein of about 8.5 kDa molecular weight, which has an ATP dependent role in the targeting of proteins for proteolytic degradation. To perform this function, the protein to be degraded is first covalently attached to the C terminus of ubiquitin, and the ubiquitinated complex is then recognized by a complex of degradative enzymes. Interestingly, ubiquitin also becomes covalently bonded to many types of pathological inclusions, which appear to be resistant to normal degradation. Therefore, ubiquitin antibodies are very useful for studies of these inclusions. For example, the neurofibrillary tangles and paired helical filaments diagnostic of Alzheimer's disease, Lewy bodies seen in Parkinson's disease, and Pick bodies found in Pick's disease are all heavily ubiquitinated and can be readily visualized with ubiquitin antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin B
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene UBB
Gene Accession No. P0CG47, P0CG49
Gene Alias AL033289; epididymis secretory protein Li 50; HEL-S-50; HUBCEP52; im:6892314; im:6892314 protein; MGC127041; polyubiquitin; polyubiquitin B; polyubiquitin-B; RPL40; Rps27a; si:ch211-202a12.3; Uba52; Ubb; Ubb2; UBC; Ubi-p63E; Ubiquitin; ubiquitin B; ubiquitin C; Ubiquitin-related; zgc:172187; zUBC
Gene Symbols UBB
Host Species Mouse
Immunogen Purified recombinant fragment of human UBB expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22187, 7314
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.