Learn More
Invitrogen™ Ub-K48 Recombinant Rabbit Monoclonal Antibody (5Y8J0)

Description
Ub-K48 Recombinant Monoclonal Antibody for ELISA, ICC/IF, Western Blot
Specifications
Specifications
| Antigen | Ub-K48 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 5Y8J0 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | UBB |
| Gene Accession No. | P0CG47 |
| Gene Alias | K48-linkage Specific Ubiquitin; Ubiquitin K48; UBK84 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.