Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ubiquitin C Recombinant Mouse Monoclonal Antibody (6G8)
GREENER_CHOICE

Catalog No. FEMA550259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550259 Supplier Invitrogen™ Supplier No. MA550259
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Ubiquitin C Recombinant Monoclonal Antibody for Flow, ELISA

This gene encodes ubiquitin, one of the most conserved proteins known. Ubiquitin is required for ATP-dependent, nonlysosomal intracellular protein degradation of abnormal proteins and normal proteins with a rapid turnover. Ubiquitin is covalently bound to proteins to be degraded, and presumably labels these proteins for degradation. Ubiquitin also binds to histone H2A in actively transcribed regions but does not cause histone H2A degradation, suggesting that ubiquitin is also involved in regulation of gene expression. This gene consists of three direct repeats of the ubiquitin coding sequence with no spacer sequence. Consequently, the protein is expressed as a polyubiquitin precursor with a final amino acid after the last repeat. Aberrant form of this protein has been noticed in patients with Alzheimer's and Down syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin C
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 6G8
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene UBC
Gene Accession No. P0CG48
Gene Alias 2700054O04Rik; AI194771; HMG20; polyubiquitin C; polyubiquitin-C; Rps27a; TI-225; Uba52; Ubb; Ubc; Ubiquitin; ubiquitin C; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2
Gene Symbols UBC
Host Species Mouse
Immunogen Recombinant Human UBC protein.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7316
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.