Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Mouse anti-Bovine, Human, APC, Clone:6C11-B3, Abnova™

Catalog No. 89333696 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-696 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-696 Supplier Abnova Corporation Supplier No. MAB16966
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against bovine Ubiquitin.

Specifications

Antigen Ubiquitin
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6C11-B3
Conjugate APC
Description polyubiquitin
Dilution ELISA
Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene LOC281370
Host Species Mouse
Immunogen Native bovine ubiquitin, conjugated to KLH.
Quantity 200 μg
Gene ID (Entrez) 281370
Target Species Bovine, Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.