Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990831
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0831 25 μL
50-199-0830 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0831 Supplier Rockland Immunochemicals Supplier No. 200401431S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum.

Ubiquitin, a highly conserved protein that has a major role in targeting cellular proteins for degradation by the 26S proteosome, is synthesized as a precursor protein consisting of either polyubiquitin chains or a single ubiquitin fused to an unrelated protein. This gene encodes a fusion protein consisting of ubiquitin at the N terminus and ribosomal protein S27a at the C terminus. When expressed in yeast, the protein is post-translationally processed, generating free ubiquitin monomer and ribosomal protein S27a. Ribosomal protein S27a is a component of the 40S subunit of the ribosome and belongs to the S27AE family of ribosomal proteins. It contains C4-type zinc finger domains and is located in the cytoplasm. Pseudogenes derived from this gene are present in the genome. As with ribosomal protein S27a, ribosomal protein L40 is also synthesized as a fusion protein with ubiquitin; similarly, ribosomal protein S30 is synthesized as a fusion protein with the ubiquitin-like protein fubi. Multiple alternatively spliced transcript variants that encode the same proteins have been identified.

Specifications

Antigen Ubiquitin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene RPS27A
Gene Accession No. P0CG47, P0CG48, P0CG49, P0CG50, P0CG51, P24051, P62979, P62982, P62983, Q63429, Q6ZWY3, Q71UM5
Gene Alias 0610006J14Rik; 1810034D23Rik; 2700054O04Rik; 40S ribosomal protein S27a; 40S ribosomal protein S27-like; 40S ribosomal protein S27-like protein; AI194771; AL033289; BOS_10808; BOS_11463; BOS_16579; BOS_18906; CEP80; Chap1; epididymis luminal protein 112; epididymis secretory protein Li 50; HEL112; HEL-S-50; HMG20; hPLIC-2; HRIHFB2157; HUBCEP52; LOC100912032; OTTHUMP00000200786; OTTHUMP00000201692; PLIC-2; polyubiquitin; polyubiquitin B; polyubiquitin C; Polyubiquitin-B; Polyubiquitin-C; ribosomal protein S27; ribosomal protein S27 like; ribosomal protein S27a; ribosomal protein S27a L homeolog; ribosomal protein S27a S homeolog; ribosomal protein S27-like; ribosomal protein S27-like protein; RPL40; RPS27A; rps27a.L; rps27a.S; Rps27l; S27A; Small ribosomal subunit protein eS27-like; Small ribosomal subunit protein eS31; TI-225; Uba52; UBA80; Ubb; Ubb2; Ubc; UBCEP1; UBCEP80; Ubiquilin 2; Ubiquitin; ubiquitin and ribosomal protein S27a; ubiquitin B; ubiquitin C; Ubiquitin carboxyl extension protein 80; ubiquitin/ribosomal protein; Ubiquitin-40S ribosomal protein S27a; ubiquitin-40S ribosomal protein S27a-like; ubiquitin-CEP80; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2; XELAEV_18028796mg
Gene Symbols RPS27A, RPS27L, UBB, UBC
Host Species Rabbit
Immunogen This purified antibody was prepared from rabbit serum after repeated immunizations with ubiquitin coupled to rabbit IgG using glutaraldehyde.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912032, 192255, 22187, 22190, 50522, 51065, 6233, 67941, 681429, 7314, 7316, 78294
Target Species Human, Multi-species, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.