Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ubiquitin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ubiquitin |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-553-910
![]() ![]() |
Proteintech Group Inc
102012AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Ubiquitin is a highly conserved globular 76 amino acid protein of about 8. 5 kDa molecular weight. It has a important role in the targeting of proteins for proteolytic degradation. Proteins to be degraded are covalently coupled to the C-terminus of ubiquitin by means of ubiquitin ligases. The ubiquitin itself is frequently also ubiquitinated, producing a polyubiquitin chain.Specifications
ubiquitin | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P0CG47, P0CG49, P0CG51 | |
192255, 22187, 7314 | |
ubiquitin Fusion Protein Ag0260 | |
Primary | |
-20°C | |
UBB |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Polyubiquitin B, UBB, ubiquitin, ubiquitin B | |
UBB | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title