Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Rabbit anti-Multi-Species, Clone: 1001C, R&D Systems™

Catalog No. A101RD
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A101RD 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A101RD Supplier R&D Systems Supplier No. A101
This item is not returnable. View return policy

Rabbit Monoclonal Antibody has been used in 2 publications

Ubiquitin Monoclonal specifically detects Ubiquitin in Multi-Species samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Ubiquitin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1001C
Conjugate Unconjugated
Dilution Western Blot 0.5 μg;/mL, Immunocytochemistry 3 μg;/mL
Formulation 0.5 mg/mL in PBS, pH 7.4, 50% glycerol, 0.09% sodium azide with 0.09% Sodium Azide
Gene Accession No. P0CG47
Gene Alias RPS27A, UBA52, UBB, UBB ubiquitin B, UBC
Gene Symbols RPS27A
Host Species Rabbit
Immunogen K48-linkage from human ubiquitin, Accession No. P0CG47
Purification Method Protein A or G purified from cell culture supernatant
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 6233.0
Test Specificity This antibody detects endogenous, human proteins containing K48-linked polyubiquitin chains in Western blots. This antibody detects purified, recombinant K48-linked polyubiquitin chains, but has no cross-reactivity to monoubiquitin or polyubiquitin of other linkages
Target Species All species
Content And Storage 12 months from date of receipt, -20°C as supplied. 3 months, -20°C under sterile conditions after opening.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.