Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant full-length human ubiquitin protein
Molecular Weight: 8.5kDa
Positive Control: Human brain from patient with Alzheimer's disease
Cellular Localization: Nuclear, cytoplasmic and membrane This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Ubiquitin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant full-length human ubiquitin protein |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Primary or Secondary | Primary |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.