Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitously expressed transcript Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6749R

Description
This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms.Specifications
Ubiquitously expressed transcript | |
Polyclonal | |
Unconjugated | |
UXT | |
0910002B17Rik; Androgen receptor trapped clone 27 protein; ART-27; BOS_25935; HSPC024; LOC100341210; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; Ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; UXT | |
Rabbit | |
Protein A | |
RUO | |
8409 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9UBK9 | |
UXT | |
KLH conjugated synthetic peptide derived from human UXT/Ubiquitously expressed transcript. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction