Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquityl-Histone H2B Mouse anti-Mouse, Human, Rat, Unconjugated, Clone: 56, MilliporeSigma™

Catalog No. 051312I25UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-131-2I25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-131-2I25UG Supplier MilliporeSigma Supplier No. 051312I25UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Ubiquityl-Histone H2B, clone 56 Antibody, Cat. No. 05-1312-I, is a highly specific mouse monoclonal antibody that targets Ubiquityl-Histone H2B and has been tested in Chromatin Immunoprecipitation (ChIP) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquityl-Histone H2B
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone 56
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Histone H2B type 1-C/E/F/G/I;Histone H2B.1 A;Histone H2B.a;H2B/al;Histone H2B.g;H2B/g;Histone H2B.h;H2B/h;Histone H2B.k;H2B/k;Histone H2B.l;H2B/l)
Gene Symbols HIST1H2BC;H2BFL;HIST1H2BE;H2BFH;HIST1H2BF;H2BFG;HIST1H2BG;H2BFA;HIST1;H2BI;H2BFK
Host Species Mouse
Immunogen A branched peptide corresponding to the ubiquitination site on human H2B.
Molecular Weight of Antigen ∽25 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 56 recognizes histone H2B when ubiquitinated on Lys120. It does not recognize H2B in its unmodified state or ubiquitin unconjugated to H2B.
Target Species Mouse, Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.