Learn More
UBL3 Mouse anti-Human, Clone: OTI1G4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807783

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Ubiquitin is a 77 amino acid protein that targets proteins for degradation by the 26S proteasome. Ubiquitin-like proteins are not directly involved in protein degradation, but appear to have many mechanistic similarities with the ubiquitin pathway. UBL3 (ubiquitin-like protein 3), also known as membrane-anchored ubiquitin-fold protein (MUB) or PNSC1, is a 117 amino acid membrane protein belonging to the ubiquitin-like family. Highly conserved between species, UBL3 contains two potential N-glycosylation sites, a potential protein kinase C phosphorylation site, and a potential C-terminal prenylation site. The gene encoding UBL3 is localized to chromosome 13q12.3.Specifications
UBL3 | |
Monoclonal | |
Unconjugated | |
UBL3 | |
HCG-1, PNSC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5412 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1G4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95164 | |
UBL3 | |
Human recombit protein fragment corresponding to amino acids 1-114 of human UBL3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.