Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562838 Supplier Invitrogen™ Supplier No. PA562838
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSNVPADMI NLRLILVSGK TKEFLFSPND SASDIAKHVY DNWPMDWEEE QVSSPNILR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Ubiquitin is a 77 amino acid protein that targets proteins for degradation by the 26S proteasome. Ubiquitin-like proteins are not directly involved in protein degradation, but appear to have many mechanistic similarities with the ubiquitin pathway. UBL3 (ubiquitin-like protein 3), also known as membrane-anchored ubiquitin-fold protein (MUB) or PNSC1, is a 117 amino acid membrane protein belonging to the ubiquitin-like family. Highly conserved between species, UBL3 contains two potential N-glycosylation sites, a potential protein kinase C phosphorylation site, and a potential C-terminal prenylation site. The gene encoding UBL3 is localized to chromosome 13q12. 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBL3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBL3
Gene Accession No. O95164
Gene Alias AW108023; HCG; HCG-1; hsMUB; Membrane-anchored ubiquitin-fold protein; mmMUB; MUB; PNSC1; Protein HCG-1; ubiquitin like 3; ubiquitin-like 3; ubiquitin-like protein 3; Ubl3; Unknown (protein for MGC:128515)
Gene Symbols UBL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBL3. Recombinant protein control fragment (Product #RP-97935).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5412
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.