Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBL4A Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
SDP

Catalog No. 502541544
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1544 30 μL
50-167-3502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1544 Supplier OriGene Supplier No. TA502447S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBL4A Monoclonal antibody specifically detects UBL4A in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen UBL4A
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2D7
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CHCHD5
Gene Accession No. Q9BSY4
Gene Alias DX254E; DXHXS254E; DXS254E; DXS254Eh; G6PD; GDX; GET5; housekeeping protein DXS254E; hypothetical protein LOC504533; MDY2; RGD1563983; TMA24; ubiquitin like 4A; ubiquitin-like 4; ubiquitin-like 4A; ubiquitin-like protein 4A; Ubiquitin-like protein GDX; UBL4; UBL4A
Host Species Mouse
Immunogen Full length human recombinant protein of human CHCHD5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84269
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.