Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBL4A Monoclonal Antibody (OTI4B6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502471S


Description
UBL4A Monoclonal antibody specifically detects UBL4A in Canine, Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
UBL4A | |
Monoclonal | |
0.19 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P40261 | |
Mouse | |
Affinity chromatography | |
RUO | |
4837, 489396 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunocytochemistry, Western Blot | |
OTI4B6 | |
Unconjugated | |
NNMT | |
DX254E; DXHXS254E; DXS254E; DXS254Eh; G6PD; GDX; GET5; housekeeping protein DXS254E; hypothetical protein LOC504533; MDY2; RGD1563983; TMA24; ubiquitin like 4A; ubiquitin-like 4; ubiquitin-like 4A; ubiquitin-like protein 4A; Ubiquitin-like protein GDX; UBL4; UBL4A | |
Full length human recombinant protein of human NNMT produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction