Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBL4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551974
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551974 Supplier Invitrogen™ Supplier No. PA551974
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSTLKQLVSE KLNVPVRQQR LLFKGKALAD GKRLSDYSIG PNSKLNLVVK PLEKVLLEEG EAQRLADSPP PQVWQLISKV LARHFSAADA SRVLEQLQRD YERSLSRLTL DDIERLASRF LHPEVTETME KG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

UBL4A is a component of the BAT3 complex, a multiprotein complex involved in the post-translational delivery of tail-anchored (TA) membrane proteins to the endoplasmic reticulum membrane. UBL4A is the TA membrane proteins, also named type II transmembrane proteins, contain a single C-terminal transmembrane region. The complex acts by facilitating TA proteins capture by ASNA1/TRC40: it is recruited to ribosomes synthesizing membrane proteins, interacts with the transmembrane region of newly released TA proteins, and transfers them to ASNA1/TRC40 for targeting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBL4A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBL4A
Gene Accession No. B2GV38, P11441, P21126
Gene Alias DX254E; DXHXS254E; DXS254E; DXS254Eh; G6PD; GDX; GET5; housekeeping protein DXS254E; hypothetical protein LOC504533; MDY2; RGD1563983; TMA24; ubiquitin like 4A; ubiquitin-like 4; ubiquitin-like 4A; ubiquitin-like protein 4A; Ubiquitin-like protein GDX; UBL4; UBL4A
Gene Symbols UBL4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBL4A. Recombinant protein control fragment (Product #RP-92186).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27643, 293864, 8266
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.