Learn More
Invitrogen™ UBL4B Monoclonal Antibody (3B2)

Description
Peptide Sequence: MFLTVKLLLG QRCSLKVSGQ ESVATLKRLV SRRLKVPEEQ QHLLFRGQLL EDDKHLSDYC IGPNASINVI MQPLEKMALK EAHQPQTQPL WHQLGLVLAK HFEPQDAKAV LQLLRQEHEE RLQKISLEHL EQLAQYLLAE EPHVEPAGER ELEAKARPQS SCDMEEKEEA AADQ.
Specifications
Specifications
| Antigen | UBL4B |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3B2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | UBL4B |
| Gene Accession No. | Q8N7F7 |
| Gene Alias | 4930522D07Rik; FLJ25690; NEWGENE_1589125; ubiquitin like 4B; ubiquitin-like 4B; ubiquitin-like protein 4B; UBL4B |
| Gene Symbols | UBL4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.