Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBL4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556716 Supplier Invitrogen™ Supplier No. PA556716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVPEEQQHLL FRGQLLEDDK HLSDYCIGPN ASINVIMQPL EKMALKEAHQ PQTQPLWHQL GLVLAKHFEP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 47%.

Ubiquitin is a 77 amino acid protein that targets proteins for degradation by the 26S proteasome. Ubiquitin-like proteins are not directly involved in protein degradation, but appear to have many mechanistic similarities with the ubiquitin pathway. UBL4B (Ubiquitin-like protein 4B) is a 174 amino acid cytoplasmic protein that contains a ubiquitin-like domain. UBL4B likely arose by retroposition during mammalian evolution from UBL4A, an X-linked intron-bearing housekeeping gene. While UBL4A is highly conserved, UBL4B has undergone rapid evolution and may have evolved new functions. Expression of UBL4B is restricted to post-meiotic germ cells in testis and ovarian tissue, where it likely functions in post-translational protein modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBL4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBL4B
Gene Accession No. Q8N7F7
Gene Alias 4930522D07Rik; FLJ25690; NEWGENE_1589125; ubiquitin like 4B; ubiquitin-like 4B; ubiquitin-like protein 4B; UBL4B
Gene Symbols UBL4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBL4B. Recombinant protein control fragment (Product #RP-94514).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.