Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBL5 Monoclonal Antibody (429CT16.3.1)
GREENER_CHOICE

Catalog No. PIMA537558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537558 Supplier Invitrogen™ Supplier No. MA537558
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBL5 Monoclonal Antibody for Western Blot

This gene encodes a member of a group of proteins similar to ubiquitin. The encoded protein is not thought to degrade proteins like ubiquitin but to affect their function through being bound to target proteins by an isopeptide bond. The gene product has been studied as a link to predisposition to obesity based on its expression in Psammomys obesus, the fat sand rat, which is an animal model for obesity studies. Variation in this gene was found to be significantly associated with some metabolic traits (PMID: 15331561) but not associated with childhood obesity (PMID: 19189687). Pseudogenes of this gene are located on chromosomes 3, 5 and 17. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBL5
Applications Western Blot
Classification Monoclonal
Clone 429CT16.3.1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide; pH 7.4
Gene UBL5
Gene Accession No. Q9BZL1, Q9EPV8
Gene Alias 1110030M22Rik; beacon; HUB1; similar to ubiquitin-like 5; testicular tissue protein Li 217; ubiquitin like 5; ubiquitin-like 5; ubiquitin-like 5 protein; ubiquitin-like protein 5; Ubl5; Unknown (protein for MGC:127118); zgc:66388
Gene Symbols UBL5
Host Species Mouse
Immunogen purified His-tagged UBL5 protein (fragment).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59286, 66177
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.