Learn More
Invitrogen™ UBL5 Monoclonal Antibody (429CT16.3.1)

Description
UBL5 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | UBL5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 429CT16.3.1 |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | UBL5 |
| Gene Accession No. | Q9BZL1, Q9EPV8 |
| Gene Alias | 1110030M22Rik; beacon; HUB1; similar to ubiquitin-like 5; testicular tissue protein Li 217; ubiquitin like 5; ubiquitin-like 5; ubiquitin-like 5 protein; ubiquitin-like protein 5; Ubl5; Unknown (protein for MGC:127118); zgc:66388 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.