Learn More
UBL7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302971AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
UBL7 (Ubiquitin Like 7) is a Protein Coding gene.Specifications
UBL7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q91W67, Q96S82 | |
Ubl7 | |
Between 330 and 380 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
UBL7 | |
BMSC-UbP, Bone marrow stromal cell ubiquitin-like protein, bone marrow stromal cell-derived ubiquitin-like, MGC14421, TCBA1, ubiquitin-like 7 (bone marrow stromal cell-derived), ubiquitin-like protein SB132 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
69459, 84993 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.