Learn More
UBL7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302970AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
UBL7 (Ubiquitin Like 7) is a Protein Coding gene.Specifications
UBL7 | |
Polyclonal | |
Unconjugated | |
UBL7 | |
BMSC-UbP, Bone marrow stromal cell ubiquitin-like protein, bone marrow stromal cell-derived ubiquitin-like, MGC14421, TCBA1, ubiquitin-like 7 (bone marrow stromal cell-derived), ubiquitin-like protein SB132 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84993 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96S82 | |
UBL7 | |
Between 1 and 50 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.