Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBLCP1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298561IR
Description
UBLCP1 Polyclonal specifically detects UBLCP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
UBLCP1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
134510 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
CPUB1, CTD phosphatase-like with ubiquitin domain 1, CTD-like phosphatase domain-containing protein, EC 3.1.3.16, FLJ25267, MGC10067, ubiquitin-like domain containing CTD phosphatase 1, ubiquitin-like domain-containing CTD phosphatase 1 | |
Produced in rabbits immunized with E. coli-derived Human UBLCP1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction