Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBLCP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110542 Supplier Invitrogen™ Supplier No. PA5110542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDIEDEVVEV ENREENLLKI SRRVKEYKVE ILNPPREGKK LLVLDVDYTL FDHRSCAETG VELMRPYLHE FLTSAYEDYD IVIWSATNMK WIEAKMKELG VSTNANYKIT FMLDSAAMIT VHTPRRGLID VKPLGVIWGK FSEFYSKKNT IMFDDIGRNF LMNPQNGLKI RPFMKAHLNR DKDKELLKLT QYLKEIAKLD DFLDLNHKYW ERYLSKKQGQ.

UBLCP1 may specifically dephosphorylate 'Ser-5' of the heptad repeats YSPTSPS in the C-terminal domain of the largest RNA polymerase II subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBLCP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UBLCP1
Gene Accession No. Q8BGR9, Q8WVY7
Gene Alias 4930527B16Rik; 8430435I17Rik; BC002236; CPUB1; CTD phosphatase-like with ubiquitin domain 1; CTD-like phosphatase domain-containing protein; fb33g09; MGC128319 protein; nuclear proteasome inhibitor UBLCP1; RGD1310386; ubiquitin like domain containing CTD phosphatase 1; ubiquitin-like domain containing CTD phosphatase 1; ubiquitin-like domain-containing CTD phosphatase 1; Ublcp1; wu:fb33g09; zgc:86634
Gene Symbols UBLCP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 99-318 of human UBLCP1 (NP_659486.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134510, 79560
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.