Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBN1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149983PCP
Description
UBN1 Polyclonal antibody specifically detects UBN1 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)Specifications
UBN1 | |
Polyclonal | |
Western Blot, Immunoprecipitation (Negative) | |
HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4 | |
The immunogen for this product maps to a region between residue 1084 and 1134 of human Ubinuclein-1 using the numbering given in entry NP_058632.2 (GeneID 29855). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
29855 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction