Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBN1 Antibody (UBN1-02), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262205AF405
Description
UBN1 Monoclonal specifically detects UBN1 in Human samples. It is validated for Western Blot.Specifications
UBN1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UBN1 | |
Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1. | |
0.1 mL | |
29855 | |
Human | |
IgG |
Western Blot | |
UBN1-02 | |
Western Blot | |
HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4 | |
Mouse | |
Protein A purified | |
Primary | |
The antibody SC-05 reacts with 80 kDa human secretory component glycoprotein (both free and bound in secretory IgA). Recognized glycoprotein is specific membrane marker of glandular carcinomas. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction