Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubn1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992913
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2913 100 μL
50-199-2914 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2913 Supplier Active Motif Supplier No. 39869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cellular senescence is a hallmark of tumor suppression and tissue aging. Senescent cells contain domains of heterochromatin, called senescence-associated heterochromatin foci (SAHF), that repress proliferation-promoting genes. The protein encoded by this gene binds to proliferation-promoting genes and is required for SAHF formation, enhancing methylation of histone H3.

Specifications

Antigen Ubn1
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene UBN1
Gene Accession No. Q9NPG3
Gene Alias 1110029L11Rik; 2610108L02Rik; AA673476; AW124741; HIRA-binding protein; Protein VT4; ubinuclein 1; Ubinuclein-1; Ubiquitously expressed nuclear protein; Ubn1; VT; VT4
Gene Symbols UBN1
Host Species Rabbit
Immunogen This Ubinuclein antibody was raised against a recombinant protein corresponding to amino acids 1-345 of human Ubn1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29855
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.