Learn More
UBN2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188441AP
Catalog No. 50-173-2258
Add to cart
Description
Description
Ubinuclein 1 (UBN1) is a ubiquitously expressed evolutionarily conserved protein which binds to proliferation-promoting genes that are repressed by formation of senescence-associated heterochromatin foci (SAHF). Ubinuclein 1 associates with various transcription factors and with histone methyltransferase activity, is indispensable for SAHF formation and appears to be a regulator of senescence. Although in most cells ubinuclein is localized to the nucleus, in cells forming tight junctions it is recruited to the cell adhesion complexes, dependently on the cell density.
Specifications
Specifications
UBN2
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZU65
KIAA2030, ubinuclein 2, UBN2
Rabbit
IgG
150 μL
-20°C
Primary
254048
Immunohistochemistry (Paraffin)
Unconjugated
UBN2
Liquid
UBN2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human