Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBOX5 Antibody (OTI1C5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274760H
Description
UBOX5 Monoclonal specifically detects UBOX5 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
UBOX5 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
UBOX5 | |
Recombinant protein fragment corresponding to amino acids 1-130 and C 419-487 of human UBOX5 (NP_955447) produced in E.coli. | |
0.1 mL | |
22888.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1C5 | |
Western Blot, Immunohistochemistry | |
KIAA0860U-box domain-containing protein 5, RING finger protein 37, RNF37Ubce7ip5, UBCE7IP5, ubiquitin conjugating enzyme 7 interacting protein 5, Ubiquitin-conjugating enzyme 7-interacting protein 5, U-box domain containing 5, UIP5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction