Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBOX5 Monoclonal Antibody (OTI5G9), TrueMAB™, OriGene
SDP

Catalog No. 501673947
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3947 100 μL
50-254-1204 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3947 Supplier OriGene Supplier No. TA502952
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.

Specifications

Antigen UBOX5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI5G9
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene UBOX5
Gene Accession No. O94941
Gene Alias RNF37, UBCE7IP5, UIP5, hUIP5
Gene Symbols UBOX5
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-130 and 419-487 of human UBOX5 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22888
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.